Assay Details
| Gene Symbol: Cxcr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022205.3 | NP_071541.2 | 1 - 2 | 80 | 102 | |
| GenBank mRNA | BC089804.1 | - | 1 - 2 | 60 | 7380432 | 102 |
| CO557829.1 | - | 1 - 2 | 80 | 102 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC089804.1,NM_022205.3 transcripts.
Set Membership: |
|

Cxcr4
Genomic Map