Assay Details
| Gene Symbol: Pigc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250183.1 | XP_006250245.1 | 1 - 2 | 75 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006250183.1 transcripts.
Set Membership: |
|

Pigc
Genomic Map