Assay Details
Gene Symbol: Rgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019336.1 | NP_062209.1 | 3 - 4 | 338 | 114 | |
XM_006249974.2 | XP_006250036.1 | 3 - 4 | 522 | 114 | ||
GenBank mRNA | BC098681.1 | - | 3 - 4 | 338 | 7385088 | 114 |
FQ234088.1 | - | 3 - 4 | 236 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
