Assay Details
Gene Symbol: Brinp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173115.1 | NP_775138.1 | 1 - 2 | 807 | 119 | |
GenBank mRNA | AB077853.1 | - | 1 - 2 | 807 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB077853.1, NM_173115.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
