Assay Details
| Gene Symbol: Rfwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025126.1 | NP_001020297.1 | 6 - 7 | 819 | 69 | |
| XM_006250113.2 | XP_006250175.1 | 13 - 14 | 2018 | 69 | ||
| XM_006250114.2 | XP_006250176.1 | 13 - 14 | 2006 | 69 | ||
| XM_006250115.2 | XP_006250177.1 | 12 - 13 | 1958 | 69 | ||
| XM_006250116.2 | XP_006250178.1 | 12 - 13 | 1946 | 69 | ||
| GenBank mRNA | BC091284.1 | - | 6 - 7 | 819 | 7304516 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Rfwd2
Genomic Map