Assay Details
| Gene Symbol: Brinp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173121.1 | NP_775144.1 | 5 - 6 | 1165 | 78 | |
| XM_006249965.2 | XP_006250027.1 | 5 - 6 | 1583 | 78 | ||
| GenBank mRNA | AB077854.1 | - | 5 - 6 | 1165 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brinp3
Genomic Map