Assay Details
| Gene Symbol: Tiprl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109667.1 | NP_001103137.1 | 1 - 2 | 153 | 71 | |
| GenBank mRNA | BC128780.1 | - | 1 - 2 | 153 | 7315719 | 71 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tiprl
Genomic Map