Assay Details
| Gene Symbol: Rnf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025667.1 | NP_001020838.1 | 1 - 2 | 198 | 129 | |
| XM_006249990.2 | XP_006250052.1 | 1 - 2 | 136 | 129 | ||
| GenBank mRNA | BC098941.1 | - | 1 - 2 | 198 | 7456372 | 129 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC098941.1, NM_001025667.1, XM_006249990.2 transcripts.
Set Membership: |
|

Rnf2
Genomic Map