Assay Details
| Gene Symbol: Dusp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022248.1 | NP_071584.1 | 4 - 5 | 691 | 112 | |
| GenBank mRNA | AF217233.1 | - | 4 - 5 | 691 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dusp12
Genomic Map