Assay Details
| Gene Symbol: Swt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105960.1 | NP_001099430.1 | 19 - 20 | 2793 | 67 | |
| XM_008769613.1 | XP_008767835.1 | 18 - 19 | 2633 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P437532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96468 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P437532 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P437532 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P437532 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P437532 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72301 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59141 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76192 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10768558 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768544 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768547 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768552 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768553 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768548 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768546 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768539 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768551 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768543 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768550 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768559 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768556 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768542 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768554 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768540 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768541 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768555 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768545 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768549 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768557 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10768538 | - | - |
| Rat Genome 230 2.0 Array | 1383076_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390868_at | - | - |
| Rat Genome 230 2.0 Array | 1383076_at | - | - |
| Rat Genome 230 2.0 Array | 1390868_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383076_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390868_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383076_at | - | - |
| Affymetrix HT Rat Focus Array | 1390868_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300011047.rn.1 | TC1300001618.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390868_PM_at | - | - |
| Rat Expression Set 230 | 1390868_at | - | - |
| Rat Expression Set 230 | 1383076_at | - | - |
| Rat Expression Set 230 | 1383076_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10768538 | - | - |
| Rat Genome U34 Set | rc_AA901203_at | - | - |
| Rat Genome U34 Set | rc_AI231480_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20907591 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372435 | - | - |

Swt1
Genomic Map