Assay Details
| Gene Symbol: Snrpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134358.1 | NP_599185.1 | 1 - 2 | 105 | 80 | |
| GenBank mRNA | BC083694.1 | - | 1 - 2 | 76 | 80 | |
| BC158686.1 | - | 1 - 2 | 105 | 8374870 | 80 | |
| FQ214448.1 | - | 1 - 2 | 80 | 80 | ||
| FQ220404.1 | - | 1 - 2 | 58 | 80 | ||
| FQ220639.1 | - | 1 - 2 | 121 | 80 | ||
| FQ220740.1 | - | 1 - 2 | 80 | |||
| FQ220961.1 | - | 1 - 2 | 106 | 80 | ||
| FQ229287.1 | - | 1 - 2 | 108 | 80 | ||
| FQ229512.1 | - | 1 - 2 | 121 | 80 | ||
| FQ230125.1 | - | 1 - 2 | 58 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC083694.1, BC158686.1, NM_134358.1 transcripts.
Set Membership: |
|

Snrpb
Genomic Map