Assay Details
| Gene Symbol: Snrpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134358.1 | NP_599185.1 | 5 - 6 | 655 | 82 | |
| GenBank mRNA | BC158686.1 | - | 5 - 6 | 655 | 8374870 | 82 |
| FQ214448.1 | - | 5 - 6 | 630 | 82 | ||
| FQ220404.1 | - | 5 - 6 | 608 | 82 | ||
| FQ220639.1 | - | 5 - 6 | 671 | 82 | ||
| FQ220740.1 | - | 5 - 6 | 656 | 82 | ||
| FQ220961.1 | - | 5 - 6 | 656 | 82 | ||
| FQ229287.1 | - | 5 - 6 | 658 | 82 | ||
| FQ229512.1 | - | 5 - 6 | 671 | 82 | ||
| FQ230125.1 | - | 5 - 6 | 608 | 82 | ||
| M29295.1 | - | 4 - 5 | 508 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Snrpb
Genomic Map