Assay Details
| Gene Symbol: Pank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106513.1 | NP_001099983.1 | 5 - 6 | 1198 | 76 | |
| XM_008762162.1 | XP_008760384.1 | 5 - 6 | 1143 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pank2
Genomic Map