Assay Details
Gene Symbol: Mall | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014182.1 | NP_001014204.1 | 1 - 2 | 142 | 60 | |
XM_008762181.1 | XP_008760403.1 | 1 - 2 | 179 | 60 | ||
GenBank mRNA | BC090079.1 | - | 1 - 2 | 142 | 7126923 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
