Assay Details
| Gene Symbol: Prg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031619.2 | NP_113807.2 | 2 - 3 | 94 | 81 | |
| GenBank mRNA | CB703061.1 | - | 1 - 2 | 89 | 81 | |
| D50568.1 | - | 2 - 3 | 94 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prg2
Genomic Map