Assay Details
| Gene Symbol: Rcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108586.1 | NP_001102056.1 | 5 - 6 | 1326 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P597431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133329 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P597431 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P597431 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P597431 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P159197 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P597431 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10847981 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847984 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861619 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847982 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10861617 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10847980 | - | - |
| Rat Genome 230 2.0 Array | 1393643_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382767_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375921_at | - | - |
| Rat Genome 230 2.0 Array | 1375921_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382767_at | - | - |
| Rat Genome 230 2.0 Array | 1393643_at | - | - |
| Affymetrix HT Rat Focus Array | 1393643_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382767_at | - | - |
| Affymetrix HT Rat Focus Array | 1375921_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375921_at | - | - |
| Affymetrix HT Rat Focus Array | 1393643_at | - | - |
| Affymetrix HT Rat Focus Array | 1382767_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300029021.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029016.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029019.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029015.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029014.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029018.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029012.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029013.rn.1 | TC0300003803.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300029020.rn.1 | TC0300003803.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393643_PM_at | - | - |
| Rat Expression Set 230 | 1393643_at | - | - |
| Rat Expression Set 230 | 1375921_at | - | - |
| Rat Expression Set 230 | 1382767_PM_at | - | - |
| Rat Expression Set 230 | 1375921_PM_at | - | - |
| Rat Expression Set 230 | 1382767_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10847980 | - | - |
| Rat Genome U34 Set | rc_AA893592_at | - | - |
| Rat Genome U34 Set | rc_AA875563_g_at | - | - |
| Rat Genome U34 Set | rc_AA875563_at | - | - |
| Rat Genome U34 Set | rc_AA944404_at | - | - |
| Rat Genome U34 Set | rc_AA851344_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22064125 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354661 | - | - |

Rcn1
Genomic Map