Assay Details
| Gene Symbol: Bahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234794.2 | XP_006234856.1 | 3 - 4 | 2304 | 66 | |
| XM_006234796.2 | XP_006234858.1 | 3 - 4 | 4105 | 66 | ||
| XM_008762115.1 | XP_008760337.1 | 3 - 4 | 1974 | 66 | ||
| XM_008762116.1 | XP_008760338.1 | 3 - 4 | 1971 | 66 | ||
| XM_008762117.1 | XP_008760339.1 | 3 - 4 | 1974 | 66 | ||
More Information
Set Membership: |
|

Bahd1
Genomic Map