Assay Details
| Gene Symbol: Cstf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077672.1 | NP_001071140.1 | 5 - 6 | 486 | 70 | |
| XM_008762067.1 | XP_008760289.1 | 6 - 7 | 502 | 70 | ||
| GenBank mRNA | BC127520.1 | - | 5 - 6 | 486 | 7110287 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|

Cstf3
Genomic Map