Assay Details
| Gene Symbol: Ube2l6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024755.1 | NP_001019926.1 | 2 - 3 | 588 | 79 | |
| GenBank mRNA | BC097364.1 | - | 2 - 3 | 588 | 7443068 | 79 |
| FQ220881.1 | - | 2 - 3 | 190 | 79 | ||
| FQ227712.1 | - | 2 - 3 | 160 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube2l6
Genomic Map