Assay Details
| Gene Symbol: Eif3j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077670.1 | NP_001071138.1 | 2 - 3 | 238 | 138 | |
| GenBank mRNA | BC127509.1 | - | 2 - 3 | 238 | 7454886 | 138 |
| FQ226401.1 | - | 2 - 3 | 187 | 138 | ||
More Information
Set Membership: |
|

Eif3j
Genomic Map