Assay Details
| Gene Symbol: Catsper2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012220.1 | NP_001012220.1 | 10 - 11 | 1484 | 68 | |
| XM_006234860.2 | XP_006234922.1 | 10 - 11 | 1410 | 68 | ||
| XM_008762187.1 | XP_008760409.1 | 10 - 11 | 1166 | 68 | ||
| GenBank mRNA | BC079422.1 | - | 10 - 11 | 1484 | 7114001 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Catsper2
Genomic Map