Assay Details
Gene Symbol: LOC654482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039174.1 | NP_001034263.1 | 2 - 3 | 198 | 78 | |
XM_006242497.2 | XP_006242559.1 | 2 - 3 | 666 | 78 | ||
GenBank mRNA | BC079425.1 | - | 2 - 3 | 198 | 7114230 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
