Assay Details
Gene Symbol: Zfp717 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766016.1 | XP_008764238.1 | 1 - 2 | 127 | 134 | |
XM_008766017.1 | XP_008764239.1 | 1 - 2 | 127 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008766016.1, XM_008766017.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
