Assay Details
| Gene Symbol: Zfp717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766011.1 | XP_008764233.1 | 10 - 11 | 1167 | 93 | |
| XM_008766012.1 | XP_008764234.1 | 10 - 11 | 1009 | 93 | ||
| XM_008766013.1 | XP_008764235.1 | 9 - 10 | 1116 | 93 | ||
| XM_008766014.1 | XP_008764236.1 | 9 - 10 | 958 | 93 | ||
| XM_008766015.1 | XP_008764237.1 | 10 - 11 | 964 | 93 | ||
| XM_008766016.1 | XP_008764238.1 | 9 - 10 | 1103 | 93 | ||
| XM_008766017.1 | XP_008764239.1 | 9 - 10 | 945 | 93 | ||
More Information
Set Membership: |
|

Zfp717
Genomic Map