Assay Details
Gene Symbol: Tyk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257347.1 | NP_001244276.1 | 19 - 20 | 3023 | 62 | |
XM_006242585.2 | XP_006242647.1 | 21 - 22 | 3334 | 62 | ||
XM_006242586.2 | XP_006242648.1 | 21 - 22 | 3293 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
