Assay Details
Gene Symbol: Ccdc67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014087.1 | NP_001014109.2 | 3 - 4 | 330 | 66 | |
XM_008765933.1 | XP_008764155.1 | 3 - 4 | 513 | 66 | ||
XM_008765934.1 | XP_008764156.1 | 3 - 4 | 446 | 66 | ||
XM_008765935.1 | XP_008764157.1 | 2 - 3 | 555 | 66 | ||
XM_008765936.1 | XP_008764158.1 | 3 - 4 | 513 | 66 | ||
XM_008765937.1 | XP_008764159.1 | 3 - 4 | 511 | 66 | ||
XM_008765938.1 | XP_008764160.1 | 3 - 4 | 511 | 66 | ||
GenBank mRNA | BC085333.1 | - | 3 - 4 | 330 | 7114511 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
