Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108995.2 | NP_001102465.2 | 2 - 3 | 175 | 110 | |
| XM_006242652.2 | XP_006242714.1 | 2 - 3 | 456 | 110 | ||
| GenBank mRNA | BC166711.1 | - | 2 - 3 | 175 | 7930337 | 110 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC166711.1,NM_001108995.2,XM_006242652.2 transcripts.
Set Membership: |
|

Zfp26
Genomic Map