Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108995.2 | NP_001102465.2 | 8 - 9 | 823 | 79 | |
| XM_006242652.2 | XP_006242714.1 | 8 - 9 | 1104 | 79 | ||
| XM_006242653.2 | XP_006242715.1 | 7 - 8 | 859 | 79 | ||
| GenBank mRNA | BC166711.1 | - | 8 - 9 | 823 | 7930337 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp26
Genomic Map