Assay Details
Gene Symbol: Prtfdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106127.1 | NP_001099597.1 | 5 - 6 | 463 | 58 | |
XM_008771860.1 | XP_008770082.1 | 5 - 6 | 474 | 58 | ||
GenBank mRNA | BC166887.1 | - | 5 - 6 | 460 | 6890546 | 58 |
BC166923.1 | - | 5 - 6 | 460 | 6919422 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
