Assay Details
| Gene Symbol: Atp5c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053825.2 | NP_446277.1 | 5 - 6 | 672 | 88 | |
| XM_006254229.2 | XP_006254291.1 | 5 - 6 | 726 | 88 | ||
| XM_006254230.2 | XP_006254292.1 | 5 - 6 | 727 | 88 | ||
| XM_006254231.2 | XP_006254293.1 | 5 - 6 | 638 | 88 | ||
| GenBank mRNA | BC059158.1 | - | 5 - 6 | 676 | 6920493 | 88 |
| FQ212651.1 | - | 5 - 6 | 670 | 88 | ||
| FQ213785.1 | - | 5 - 6 | 672 | 88 | ||
| FQ214863.1 | - | 5 - 6 | 726 | 88 | ||
| FQ215205.1 | - | 5 - 6 | 670 | 88 | ||
| FQ215286.1 | - | 5 - 6 | 645 | 88 | ||
| FQ215351.1 | - | 5 - 6 | 670 | 88 | ||
| FQ215426.1 | - | 5 - 6 | 669 | 88 | ||
| FQ215847.1 | - | 5 - 6 | 670 | 88 | ||
| FQ216198.1 | - | 5 - 6 | 666 | 88 | ||
| FQ216285.1 | - | 5 - 6 | 660 | 88 | ||
| FQ216490.1 | - | 5 - 6 | 670 | 88 | ||
| FQ216539.1 | - | 5 - 6 | 654 | 88 | ||
| FQ216647.1 | - | 5 - 6 | 672 | 88 | ||
| FQ216736.1 | - | 5 - 6 | 670 | 88 | ||
| FQ218995.1 | - | 5 - 6 | 672 | 88 | ||
| FQ219484.1 | - | 5 - 6 | 670 | 88 | ||
| FQ219563.1 | - | 5 - 6 | 670 | 88 | ||
| FQ219689.1 | - | 5 - 6 | 672 | 88 | ||
| FQ220076.1 | - | 5 - 6 | 670 | 88 | ||
| FQ220217.1 | - | 5 - 6 | 672 | 88 | ||
| FQ220355.1 | - | 5 - 6 | 670 | 88 | ||
| FQ224939.1 | - | 5 - 6 | 670 | 88 | ||
| FQ229987.1 | - | 5 - 6 | 693 | 88 | ||
| FQ234981.1 | - | 5 - 6 | 670 | 88 | ||
| L19927.1 | - | 3 - 4 | 550 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5c1
Genomic Map