Assay Details
| Gene Symbol: Atp5c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053825.2 | NP_446277.1 | 7 - 8 | 929 | 104 | |
| XM_006254231.2 | XP_006254293.1 | 7 - 8 | 895 | 104 | ||
| GenBank mRNA | BC059158.1 | - | 7 - 8 | 933 | 6920493 | 104 |
| FQ212651.1 | - | 7 - 8 | 927 | 104 | ||
| FQ213852.1 | - | 7 - 8 | 929 | 104 | ||
| FQ218995.1 | - | 7 - 8 | 929 | 104 | ||
| FQ219484.1 | - | 7 - 8 | 927 | 104 | ||
| FQ219689.1 | - | 7 - 8 | 929 | 104 | ||
| FQ229987.1 | - | 7 - 8 | 950 | 104 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5c1
Genomic Map