Assay Details
| Gene Symbol: Akr1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138510.1 | NP_612519.1 | 2 - 3 | 300 | 78 | |
| GenBank mRNA | BC086579.1 | - | 2 - 3 | 299 | 78 | |
| BC100248.1 | - | 2 - 3 | 294 | 7307214 | 78 | |
| D14424.1 | - | 2 - 3 | 285 | 78 | ||
| L32601.1 | - | 2 - 3 | 300 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akr1c3
Genomic Map