Assay Details
| Gene Symbol: Akr1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013057.1 | NP_001013075.1 | 2 - 3 | 278 | 65 | |
| XM_006254183.2 | XP_006254245.1 | 2 - 3 | 290 | 65 | ||
| XM_006254184.2 | XP_006254246.1 | 2 - 3 | 269 | 65 | ||
| XM_006254185.2 | XP_006254247.1 | 2 - 3 | 290 | 65 | ||
| GenBank mRNA | BC078957.1 | - | 2 - 3 | 278 | 7110806 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Akr1c2
Genomic Map