Assay Details
| Gene Symbol: Akr1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254183.2 | XP_006254245.1 | 8 - 9 | 1057 | 89 | |
| XM_006254184.2 | XP_006254246.1 | 8 - 9 | 1036 | 89 | ||
| XM_006254185.2 | XP_006254247.1 | 7 - 8 | 979 | 89 | ||
More Information
Set Membership: |
|

Akr1c2
Genomic Map