Assay Details
| Gene Symbol: Pter | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022224.2 | NP_071560.2 | 2 - 3 | 494 | 82 | |
| XM_006254290.2 | XP_006254352.2 | 2 - 3 | 3763 | 82 | ||
| XM_006254291.2 | XP_006254353.1 | 2 - 3 | 581 | 82 | ||
| GenBank mRNA | BC078833.1 | - | 2 - 3 | 494 | 7110694 | 82 |
More Information
Set Membership: |
|

Pter
Genomic Map