Assay Details
Gene Symbol: Ucma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254265.1 | XP_006254327.1 | 1 - 2 | 660 | 96 | |
GenBank mRNA | EU022754.1 | - | 1 - 2 | 95 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
