Assay Details
Gene Symbol: Ucma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254265.1 | XP_006254327.1 | 2 - 3 | 720 | 93 | |
GenBank mRNA | EU022754.1 | - | 2 - 3 | 155 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
