Assay Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170343.1 | NP_001163814.1 | 3 - 4 | 209 | 77 | |
| XM_006254257.2 | XP_006254319.1 | 3 - 4 | 309 | 77 | ||
| GenBank mRNA | CK470805.1 | - | 3 - 4 | 232 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec61a2
Genomic Map