Assay Details
| Gene Symbol: Spag6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106125.1 | NP_001099595.1 | 9 - 10 | 1320 | 111 | |
| XM_006254307.2 | XP_006254369.1 | 10 - 11 | 5234 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spag6l
Genomic Map