Assay Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254331.2 | XP_006254393.1 | 6 - 7 | 2280 | 86 | |
| XM_006254332.2 | XP_006254394.1 | 6 - 7 | 1466 | 86 | ||
| XM_008771904.1 | XP_008770126.1 | 9 - 10 | 2217 | 86 | ||
| XM_008771905.1 | XP_008770127.1 | 9 - 10 | 2217 | 86 | ||
| XM_008771906.1 | XP_008770128.1 | 8 - 9 | 2112 | 86 | ||
| XM_008771907.1 | XP_008770129.1 | 8 - 9 | 2112 | 86 | ||
| XM_008771908.1 | XP_008770130.1 | 9 - 10 | 2217 | 86 | ||
| XM_008771909.1 | XP_008770131.1 | 9 - 10 | 1839 | 86 | ||
| XM_008771910.1 | XP_008770132.1 | 9 - 10 | 2219 | 86 | ||
| XM_008771911.1 | XP_008770133.1 | 8 - 9 | 2113 | 86 | ||
| XM_008771912.1 | XP_008770134.1 | 9 - 10 | 2219 | 86 | ||
| XM_008771913.1 | XP_008770135.1 | 8 - 9 | 2112 | 86 | ||
More Information
Set Membership: |
|

Etl4
Genomic Map