Assay Details
Gene Symbol: Tusc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004212.1 | NP_001004212.1 | 1 - 2 | 327 | 80 | |
XM_008771294.1 | XP_008769516.1 | 1 - 2 | 345 | 80 | ||
GenBank mRNA | BC079193.1 | - | 1 - 2 | 327 | 7131345 | 80 |
FQ213307.1 | - | 1 - 2 | 345 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
