Assay Details
Gene Symbol: Snx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253168.1 | XP_006253230.1 | 9 - 10 | 1457 | 68 | |
XM_224863.6 | XP_224863.5 | 10 - 11 | 1619 | 68 | ||
GenBank mRNA | BC086510.1 | - | 4 - 5 | 629 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
