Assay Details
| Gene Symbol: Abhd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108866.1 | NP_001102336.1 | 2 - 3 | 175 | 125 | |
| XM_006251995.2 | XP_006252057.1 | 2 - 3 | 380 | 125 | ||
| XM_006251996.2 | XP_006252058.1 | 2 - 3 | 171 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006251995.2, XM_006251996.2 transcripts.
Set Membership: |
|

Abhd4
Genomic Map