Assay Details
Gene Symbol: Pabpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135008.1 | NP_001128480.1 | 2 - 3 | 466 | 104 | |
XM_008770659.1 | XP_008768881.1 | 3 - 4 | 2121 | 104 | ||
GenBank mRNA | U94858.1 | - | 2 - 3 | 169 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
