Assay Details
Gene Symbol: Lrrc16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014138.2 | NP_001014160.2 | 27 - 28 | 2490 | 58 | |
XM_008770681.1 | XP_008768903.1 | 27 - 28 | 3299 | 58 | ||
XM_008770682.1 | XP_008768904.1 | 27 - 28 | 3299 | 58 | ||
XM_008770683.1 | XP_008768905.1 | 27 - 28 | 3299 | 58 | ||
XM_008770684.1 | XP_008768906.1 | 27 - 28 | 3299 | 58 | ||
XM_008770685.1 | XP_008768907.1 | 27 - 28 | 3299 | 58 | ||
XM_008770686.1 | XP_008768908.1 | 27 - 28 | 3299 | 58 | ||
XM_008770687.1 | XP_008768909.1 | 25 - 26 | 2333 | 58 | ||
GenBank mRNA | BC083918.1 | - | 3 - 4 | 431 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
