Assay Details
Gene Symbol: Cideb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108869.1 | NP_001102339.1 | 1 - 2 | 212 | 86 | |
XM_006252002.2 | XP_006252064.1 | 1 - 2 | 207 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
