Assay Details
| Gene Symbol: Ndufs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025146.1 | NP_001020317.1 | 1 - 2 | 102 | 66 | |
| GenBank mRNA | BC083729.1 | - | 1 - 2 | 102 | 7191329 | 66 |
| BC127458.1 | - | 1 - 2 | 102 | 7193742 | 66 | |
| FQ212751.1 | - | 1 - 2 | 119 | 66 | ||
| FQ214408.1 | - | 1 - 2 | 119 | 66 | ||
| FQ216263.1 | - | 1 - 2 | 102 | 66 | ||
| FQ223158.1 | - | 1 - 2 | 119 | 66 | ||
| FQ223379.1 | - | 1 - 2 | 119 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs4
Genomic Map