Assay Details
| Gene Symbol: Il6st | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008725.3 | NP_001008725.2 | 14 - 15 | 2037 | 133 | |
| XM_006231928.2 | XP_006231990.1 | 16 - 17 | 2290 | 133 | ||
| XM_008760736.1 | XP_008758958.1 | 16 - 17 | 3135 | 133 | ||
| GenBank mRNA | AY310138.1 | - | 1986 | 133 | ||
| JQ013730.1 | - | 14 - 15 | 2388 | 133 | ||
| M92340.1 | - | 14 - 15 | 2071 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycogen metabolic process
triglyceride mobilization
signal transduction
response to nutrient
positive regulation of cell proliferation
regulation of Notch signaling pathway
positive regulation of smooth muscle cell migration
cytokine-mediated signaling pathway
response to cytokine
interleukin-11-mediated signaling pathway
oncostatin-M-mediated signaling pathway
positive regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of osteoblast differentiation
regulation of hormone secretion
positive regulation of astrocyte differentiation
leukemia inhibitory factor signaling pathway
negative regulation of cytosolic calcium ion concentration
interleukin-6-mediated signaling pathway
negative regulation of interleukin-6-mediated signaling pathway
interleukin-27-mediated signaling pathway
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
interleukin-6 receptor activity
interleukin-11 receptor activity
leukemia inhibitory factor receptor activity
oncostatin-M receptor activity
ciliary neurotrophic factor receptor binding
interleukin-6 receptor binding
growth factor binding
interleukin-11 binding
interleukin-6 binding
protein heterodimerization activity
glycogen metabolic process
triglyceride mobilization
signal transduction
response to nutrient
positive regulation of cell proliferation
regulation of Notch signaling pathway
positive regulation of smooth muscle cell migration
cytokine-mediated signaling pathway
response to cytokine
interleukin-11-mediated signaling pathway
oncostatin-M-mediated signaling pathway
positive regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of osteoblast differentiation
regulation of hormone secretion
positive regulation of astrocyte differentiation
leukemia inhibitory factor signaling pathway
negative regulation of cytosolic calcium ion concentration
interleukin-6-mediated signaling pathway
negative regulation of interleukin-6-mediated signaling pathway
interleukin-27-mediated signaling pathway
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
interleukin-6 receptor activity
interleukin-11 receptor activity
leukemia inhibitory factor receptor activity
oncostatin-M receptor activity
ciliary neurotrophic factor receptor binding
interleukin-6 receptor binding
growth factor binding
interleukin-11 binding
interleukin-6 binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P084300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202637 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25228 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084300 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P705481 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084300 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P705481 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P084300 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P705481 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P363090 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700769137 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86350 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17750 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66080 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700769137 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10813008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813021 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813026 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813024 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813014 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813022 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813009 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10813007 | - | - |
| Rat Genome 230 2.0 Array | 1370957_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370957_at | - | - |
| Rat Genome 230 2.0 Array | 1371926_at | - | - |
| Rat Genome 230 2.0 Array | 1371926_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383489_at | - | - |
| Affymetrix HT Rat Focus Array | 1370957_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373140_at | - | - |
| Affymetrix HT Rat Focus Array | 1373140_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371926_at | - | - |
| Affymetrix HT Rat Focus Array | 1370957_at | - | - |
| Affymetrix HT Rat Focus Array | 1383489_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371926_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200002290.rn.1 | TC0200000390.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200002288.rn.1 | TC0200000390.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200002289.rn.1 | TC0200000390.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200002291.rn.1 | TC0200000390.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200002292.rn.1 | TC0200000390.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200002293.rn.1 | TC0200000390.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370957_PM_at | - | - |
| Rat Expression Set 230 | 1370957_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813007 | - | - |
| Rat Genome U34 Set | M92340_at | - | - |
| Rat Genome U34 Set | rc_AI044396_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22305984 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376855 | - | - |

Il6st
Genomic Map