Assay Details
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134415.2 | NP_599242.2 | 1 - 2 | 102 | 123 | |
GenBank mRNA | AF158379.1 | - | 1 - 2 | 110 | 123 | |
BC098657.1 | - | 1 - 2 | 93 | 7378282 | 123 | |
CR473148.1 | - | 1 - 2 | 123 | |||
DQ354588.1 | - | 1 - 2 | 92 | 123 | ||
FQ212851.1 | - | 1 - 2 | 114 | 123 | ||
FQ214845.1 | - | 1 - 2 | 121 | 123 | ||
FQ217534.1 | - | 1 - 2 | 115 | 123 | ||
FQ225571.1 | - | 1 - 2 | 115 | 123 | ||
FQ226635.1 | - | 1 - 2 | 129 | 123 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AF158379.1,BC098657.1,DQ354588.1,NM_134415.2 transcripts.
This assay is in the 5'UTR region of the AF158379.1,BC098657.1,DQ354588.1,NM_134415.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
