Assay Details
Gene Symbol: Nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134415.2 | NP_599242.2 | 2 - 3 | 294 | 100 | |
GenBank mRNA | AF158379.1 | - | 2 - 3 | 302 | 100 | |
BC098657.1 | - | 2 - 3 | 285 | 7378282 | 100 | |
DQ354588.1 | - | 2 - 3 | 284 | 100 | ||
FQ225571.1 | - | 2 - 3 | 307 | 100 | ||
FQ226635.1 | - | 2 - 3 | 321 | 100 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
